Assay Details
Target Gene Details
Entrez Gene ID: | 81790 |
Gene Name: | ring finger protein 170 |
Gene Aliases: |
ADSA, SNAX1 |
Location: |
Chr.8:42849637-42897288 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF170 | NM_001160223.1 | 7 | 1295 | NP_001153695.1 |
| NM_001160224.1 | NP_001153696.1 | |||
| NM_001160225.1 | 7 | 944 | NP_001153697.1 | |
| NM_030954.3 | 7 | 1065 | NP_112216.3 | |
| NR_027668.1 | 6 | 1221 | ||
| NR_027669.1 | 6 | 991 | ||
| XM_006716404.2 | 8 | 1052 | XP_006716467.1 | |
| XM_006716405.2 | 5 | 811 | XP_006716468.1 | |
| XM_011544666.2 | 7 | 849 | XP_011542968.1 | |
| XM_017013881.1 | 8 | 992 | XP_016869370.1 | |
| XM_017013882.1 | 6 | 837 | XP_016869371.1 | |
| AF209504.1 | 2 | 402 | AAM92891.1 | |
| AK027748.1 | 7 | 981 | BAB55340.1 | |
| AK090864.1 | 6 | 1154 | BAC03534.1 | |
| AK095625.1 | 1 | 493 | ||
| AK225086.1 | 7 | 945 | ||
| AK225568.1 | 7 | 961 | ||
| AL136620.1 | 7 | 949 | CAB66555.1 | |
| BC013422.1 | 6 | 856 | AAH13422.1 | |
| BC032393.1 | AAH32393.1 | |||
| BC039461.1 | 7 | 1052 | AAH39461.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483102 | Chr.8:42722031 - 42876093 on Build GRCh38 | Loss |
|
| nsv6171 | Chr.8:42853461 - 42887332 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map