Assay Details
Target Gene Details
Entrez Gene ID: | 10992 |
Gene Name: | splicing factor 3b subunit 2 |
Gene Aliases: |
Cus1, SAP145, SF3B145, SF3b1, SF3b150 |
Location: |
Chr.11:66051772-66068911 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SF3B2 | NM_006842.2 | 13 | 1525 | NP_006833.2 |
| XM_005273726.3 | 13 | 2095 | XP_005273783.1 | |
| XM_011544740.2 | 13 | 2095 | XP_011543042.1 | |
| XM_017017144.1 | 13 | 2092 | XP_016872633.1 | |
| AK290850.1 | 13 | 1509 | ||
| AK300016.1 | 7 | 799 | ||
| BC000401.2 | 13 | 1484 | AAH00401.2 | |
| BC007610.1 | 6 | 708 | AAH07610.1 | |
| BC053577.1 | 13 | 1492 | AAH53577.1 | |
| BX537771.1 | 11 | 1198 | CAD97834.1 | |
| U41371.1 | 13 | 1464 | AAA97461.1 |
More Information
Set Membership: |
|

Genomic Map