Assay Details
Target Gene Details
Entrez Gene ID: | 9183 |
Gene Name: | zw10 kinetochore protein |
Gene Aliases: |
HZW10, KNTC1AP |
Location: |
Chr.11:113733183-113773763 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZW10 | NM_004724.3 | 16 | 2419 | NP_004715.1 |
| XM_017018558.1 | 15 | 2167 | XP_016874047.1 | |
| XM_017018559.1 | 16 | 2375 | XP_016874048.1 | |
| AK300746.1 | 8 | 894 | ||
| AK303871.1 | 15 | 2098 | ||
| AK312925.1 | ||||
| BC128264.1 | 16 | 2298 | ||
| BQ773926.1 | 1 | 546 | ||
| DB506745.2 | 2 | 126 | ||
| U54996.1 | 16 | 2345 | AAB88237.1 |
More Information
Set Membership: |
|

Genomic Map