Assay Details
Target Gene Details
Entrez Gene ID: | 55731 |
Gene Name: | family with sequence similarity 222 member B |
Gene Aliases: |
C17orf63 |
Location: |
Chr.17:28755978-28855036 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM222B | NM_001077498.2 | 3 | 3125 | NP_001070966.1 |
| NM_001288631.1 | 4 | 3296 | NP_001275560.1 | |
| NM_001288632.1 | 5 | 3207 | NP_001275561.1 | |
| NM_001288633.1 | 4 | 3201 | NP_001275562.1 | |
| NM_001288634.1 | 3 | 3194 | NP_001275563.1 | |
| NM_001288635.1 | 4 | 3173 | NP_001275564.1 | |
| NM_001288636.1 | 6 | 3197 | NP_001275565.1 | |
| NM_001288637.1 | 5 | 3163 | NP_001275566.1 | |
| NM_001288638.1 | 5 | 3095 | NP_001275567.1 | |
| NM_001288639.1 | 4 | 3047 | NP_001275568.1 | |
| NM_001288640.1 | 3 | 3013 | NP_001275569.1 | |
| NM_018182.3 | 4 | 3159 | NP_060652.2 | |
| XM_011524986.2 | 3 | 3090 | XP_011523288.1 | |
| XM_011524987.1 | 4 | 2925 | XP_011523289.1 | |
| XM_011524988.2 | 3 | 3141 | XP_011523290.1 | |
| XM_017024831.1 | 4 | 3249 | XP_016880320.1 | |
| XM_017024832.1 | 3 | 3514 | XP_016880321.1 | |
| XM_017024833.1 | 3 | 3054 | XP_016880322.1 | |
| XM_017024834.1 | 3 | 3252 | XP_016880323.1 | |
| XM_017024835.1 | 4 | 3451 | XP_016880324.1 | |
| AK001562.1 | 4 | 3110 | BAA91757.1 | |
| CB156081.1 | 1 | 386 |

Genomic Map