Assay Details
Target Gene Details
Entrez Gene ID: | 143384 |
Gene Name: | CDK2 associated cullin domain 1 |
Gene Aliases: |
C10orf46, CAC1 |
Location: |
Chr.10:118680982-118755246 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACUL1 | NM_153810.4 | 9 | 3591 | NP_722517.3 |
| AK097728.1 | BAC05153.1 | |||
| AK301182.1 | ||||
| AY743663.1 | 9 | 3592 | AAU89709.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832006 | Chr.10:118557873 - 118759411 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map