Assay Details
Target Gene Details
Entrez Gene ID: | 7850 |
Gene Name: | interleukin 1 receptor type 2 |
Gene Aliases: |
CD121b, CDw121b, IL-1R-2, IL-1RT-2, IL-1RT2, IL1R2c, IL1RB |
Location: |
Chr.2:101991805-102028544 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1R2 | NM_001261419.1 | 3 | 505 | NP_001248348.1 |
| NM_004633.3 | 3 | 505 | NP_004624.1 | |
| NR_048564.1 | 3 | 492 | ||
| XM_006712734.3 | 3 | 529 | XP_006712797.1 | |
| XM_006712736.3 | 3 | 566 | XP_006712799.1 | |
| XM_011511801.2 | 3 | 2865 | XP_011510103.1 | |
| XM_011511803.2 | 3 | 2444 | XP_011510105.1 | |
| XM_011511804.2 | 3 | 558 | XP_011510106.1 | |
| XM_011511805.2 | 3 | 1331 | XP_011510107.1 | |
| XM_011511807.1 | 3 | 2865 | XP_011510109.1 | |
| XM_017004889.1 | 2 | 415 | XP_016860378.1 | |
| AK130412.1 | 3 | 389 | ||
| AK309161.1 | 2 | 261 | ||
| BC027733.2 | 3 | 459 | AAH27733.1 | |
| BC039031.1 | 3 | 505 | AAH39031.1 | |
| DC380050.1 | 3 | 493 | ||
| FJ200480.1 | 2 | 116 | ||
| U64094.1 | 2 | 337 | AAB05878.1 | |
| U74649.1 | 2 | 311 | AAD00242.1 | |
| X59770.1 | 2 | 337 | CAA42441.1 |
More Information
Set Membership: |
|

Genomic Map