Assay Details
Target Gene Details
Entrez Gene ID: | 80818 |
Gene Name: | zinc finger protein 436 |
Gene Aliases: |
ZNF, Zfp46 |
Location: |
Chr.1:23359448-23369864 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF436 | NM_001077195.1 | 4 | 2826 | NP_001070663.1 |
| NM_030634.2 | 3 | 2648 | NP_085137.1 | |
| XM_011542201.1 | 3 | 2730 | XP_011540503.1 | |
| AB051497.1 | 4 | 2404 | ||
| AK092364.1 | 4 | 2096 | BAC03871.1 | |
| AL834485.1 | 3 | 2648 | CAD39143.1 | |
| BC056400.1 | 3 | 2559 | AAH56400.1 |
More Information
Set Membership: |
|

Genomic Map