Assay Details
Target Gene Details
Entrez Gene ID: | 56907 |
Gene Name: | spire type actin nucleation factor 1 |
Gene Aliases: |
Spir-1 |
Location: |
Chr.18:12446512-12658102 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPIRE1 | NM_001128626.1 | 17 | 2326 | NP_001122098.1 |
| NM_001128627.1 | 16 | 1982 | NP_001122099.1 | |
| NM_020148.2 | 16 | 2284 | NP_064533.3 | |
| XM_005258122.4 | 17 | 2641 | XP_005258179.1 | |
| XM_011525701.1 | 18 | 2689 | XP_011524003.1 | |
| XM_011525702.1 | 18 | 2201 | XP_011524004.1 | |
| AB032961.2 | 17 | 2378 | ||
| AJ277587.1 | 14 | 1871 | CAB96370.1 | |
| AK025433.1 | 1 | 240 | ||
| AK290180.1 | 14 | 1842 | ||
| AK301711.1 | 15 | 1751 | ||
| AL833817.1 | 16 | 1986 | CAD38680.1 | |
| BC016825.1 | 4 | 483 | AAH16825.1 | |
| BC115005.1 | 16 | 2109 | AAI15006.1 | |
| BC125206.1 | 15 | 1840 | ||
| BC125207.1 | 14 | 1798 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv978607 | Chr.18:12449631 - 12451711 on Build GRCh38 | Duplication |
|
| nsv1060808 | Chr.18:12302793 - 12514418 on Build GRCh38 | Gain |
|
| nsv576496 | Chr.18:12247873 - 12490503 on Build GRCh38 | Gain |
|
| nsv1063125 | Chr.18:12046893 - 12918252 on Build GRCh38 | Loss |
|
| esv2758714 | Chr.18:12240682 - 12499526 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map