Assay Details
Target Gene Details
Entrez Gene ID: | 79931 |
Gene Name: | TNFAIP3 interacting protein 3 |
Gene Aliases: |
ABIN-3, LIND |
Location: |
Chr.4:121127161-121227609 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNIP3 | NM_001128843.2 | 13 | 1672 | NP_001122315.2 |
| NM_001244764.1 | 13 | 1712 | NP_001231693.1 | |
| NM_024873.5 | 11 | 1678 | NP_079149.3 | |
| XM_011532256.2 | 15 | 2006 | XP_011530558.1 | |
| XM_011532257.2 | 14 | 1945 | XP_011530559.1 | |
| XM_017008619.1 | 16 | 2107 | XP_016864108.1 | |
| XM_017008620.1 | 16 | 1970 | XP_016864109.1 | |
| XM_017008621.1 | 15 | 2017 | XP_016864110.1 | |
| XM_017008622.1 | 15 | 2270 | XP_016864111.1 | |
| XM_017008623.1 | 15 | 1880 | XP_016864112.1 | |
| XM_017008624.1 | 14 | 1825 | XP_016864113.1 | |
| XM_017008625.1 | 13 | 2358 | XP_016864114.1 | |
| AF277289.1 | 11 | 1667 | AAL02151.1 | |
| AJ320534.1 | 11 | 1666 | CAC85929.1 | |
| AK024815.1 | 9 | 1279 | BAB15018.1 | |
| CD367219.1 | 1 | 123 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4492 | Chr.4:121105502 - 121138401 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map