Assay Details
Target Gene Details
Entrez Gene ID: | 132321 |
Gene Name: | chromosome 4 open reading frame 33 |
Gene Aliases: |
- |
Location: |
Chr.4:129093674-129112688 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C4orf33 | NM_001099783.1 | 6 | 663 | NP_001093253.1 |
| NM_173487.2 | 6 | 935 | NP_775758.2 | |
| XM_005262734.4 | 6 | 930 | XP_005262791.1 | |
| AK091022.1 | 6 | 924 | BAC03568.1 | |
| BC016358.2 | 2 | 276 | AAH16358.1 | |
| BC032582.1 | 6 | 866 | AAH32582.1 | |
| BX538164.1 | 6 | 876 | CAD98044.1 |
More Information
Set Membership: |
|

Genomic Map