Assay Details
Target Gene Details
Entrez Gene ID: | 4670 |
Gene Name: | heterogeneous nuclear ribonucleoprotein M |
Gene Aliases: |
CEAR, HNRNPM4, HNRPM, HNRPM4, HTGR1, NAGR1, hnRNP M |
Location: |
Chr.19:8444575-8489118 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPM | NM_001297418.1 | 2 | 367 | NP_001284347.1 |
| NM_005968.4 | 2 | 276 | NP_005959.2 | |
| NM_031203.3 | 2 | 276 | NP_112480.2 | |
| XM_005272478.2 | 2 | 620 | XP_005272535.1 | |
| XM_005272479.2 | 2 | 620 | XP_005272536.1 | |
| XM_005272480.2 | 2 | 620 | XP_005272537.1 | |
| XM_005272481.1 | 2 | 275 | XP_005272538.1 | |
| XM_005272483.2 | 2 | 257 | XP_005272540.1 | |
| XM_017026823.1 | 2 | 620 | XP_016882312.1 | |
| XM_017026824.1 | 2 | 620 | XP_016882313.1 | |
| XM_017026825.1 | 2 | 620 | XP_016882314.1 | |
| XM_017026826.1 | 2 | 220 | XP_016882315.1 | |
| XM_017026827.1 | 2 | 275 | XP_016882316.1 | |
| XM_017026828.1 | 2 | 224 | XP_016882317.1 | |
| XM_017026829.1 | 2 | 275 | XP_016882318.1 | |
| XM_017026830.1 | 2 | 275 | XP_016882319.1 | |
| AB209733.1 | 2 | 195 | BAD92970.1 | |
| AF061832.1 | 2 | 426 | AAC16002.1 | |
| BC000138.2 | 2 | 237 | AAH00138.2 | |
| BC019580.1 | 2 | 205 | AAH19580.1 | |
| DB481659.1 | 2 | 276 | ||
| HY007201.1 | 2 | 368 | ||
| L03532.1 | 2 | 204 | AAA36192.1 | |
| L32611.1 | 2 | 205 | AAL31359.1 |
More Information
Set Membership: |
|

Genomic Map