Assay Details
Target Gene Details
Entrez Gene ID: | 115827 |
Gene Name: | RAB3C, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.5:58582188-58859394 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB3C | NM_001317915.1 | 2 | 264 | NP_001304844.1 |
| NM_138453.3 | 2 | 226 | NP_612462.1 | |
| AY026936.1 | 2 | 184 | AAK08968.1 | |
| BC013033.2 | 2 | 158 | AAH13033.1 | |
| BX375794.2 | 1 | 193 | ||
| BX394661.2 | 2 | 236 | ||
| DB485923.1 | 2 | 230 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830311 | Chr.5:58616719 - 58809809 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map