Assay Details
Target Gene Details
Entrez Gene ID: | 143684 |
Gene Name: | family with sequence similarity 76 member B |
Gene Aliases: |
- |
Location: |
Chr.11:95768942-95790396 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM76B | NM_144664.4 | 10 | 2636 | NP_653265.3 |
| XM_005273775.4 | 10 | 2975 | XP_005273832.1 | |
| XM_005273776.4 | 10 | 3019 | XP_005273833.1 | |
| XM_011542610.2 | 11 | 2410 | XP_011540912.1 | |
| XM_011542611.2 | 10 | 2999 | XP_011540913.1 | |
| XM_011542612.1 | 10 | 2500 | XP_011540914.1 | |
| XM_011542613.1 | 10 | 2393 | XP_011540915.1 | |
| BC026013.2 | AAH26013.1 | |||
| BC028727.1 | 11 | 2526 | AAH28727.1 | |
| BX647586.1 | 10 | 2345 | CAI46088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832235 | Chr.11:95753612 - 95935071 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map