Assay Details
Target Gene Details
Entrez Gene ID: | 23603 |
Gene Name: | coronin 1C |
Gene Aliases: |
HCRNN4 |
Location: |
Chr.12:108645109-108731550 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CORO1C | NM_001105237.2 | 10 | 1418 | NP_001098707.1 |
| NM_001276471.1 | 10 | 1436 | NP_001263400.1 | |
| NM_014325.3 | 10 | 1337 | NP_055140.1 | |
| XM_011538125.1 | 6 | 661 | XP_011536427.1 | |
| XM_017019121.1 | 11 | 2166 | XP_016874610.1 | |
| XM_017019122.1 | 10 | 1743 | XP_016874611.1 | |
| XM_017019123.1 | 11 | 1796 | XP_016874612.1 | |
| AB030656.1 | 10 | 1306 | BAA83077.1 | |
| AB209909.1 | 10 | 1292 | BAD93146.1 | |
| AK023205.1 | 10 | 1307 | ||
| AK096363.1 | 10 | 1436 | ||
| AK223073.1 | 10 | 1305 | BAD96793.1 | |
| AK297463.1 | 10 | 1170 | ||
| AK304839.1 | 10 | 1073 | ||
| AK311315.1 | 4 | 262 | ||
| AK316065.1 | 10 | 1133 | ||
| AK316504.1 | 9 | 1058 | ||
| AL162070.1 | 10 | 1275 | CAB82406.1 | |
| AM849477.1 | 10 | 1311 | ||
| AM849478.1 | 10 | 1418 | ||
| BC002342.2 | 10 | 1291 | AAH02342.1 |
More Information
Set Membership: |
|

Genomic Map