Assay Details
Target Gene Details
Entrez Gene ID: | 2274 |
Gene Name: | four and a half LIM domains 2 |
Gene Aliases: |
AAG11, DRAL, FHL-2, SLIM-3, SLIM3 |
Location: |
Chr.2:105360826-105438773 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FHL2 | NM_001039492.2 | 7 | 1282 | NP_001034581.1 |
| NM_001318894.1 | 6 | 1528 | NP_001305823.1 | |
| NM_001318895.1 | 7 | 1516 | NP_001305824.1 | |
| NM_001318896.1 | 6 | 1231 | NP_001305825.1 | |
| NM_001318897.1 | 5 | 1282 | NP_001305826.1 | |
| NM_001318898.1 | 6 | 1341 | NP_001305827.1 | |
| NM_001318899.1 | 8 | 1404 | NP_001305828.1 | |
| NM_001450.3 | 6 | 1465 | NP_001441.4 | |
| NM_201555.1 | 8 | 1637 | NP_963849.1 | |
| NM_201557.3 | 6 | 1457 | NP_963851.2 | |
| XM_011510798.2 | 7 | 1264 | XP_011509100.1 | |
| AA758053.1 | 1 | 271 | ||
| AB158503.1 | 5 | 1004 | BAD69710.1 | |
| AK097756.1 | 1 | 1868 | ||
| BC012742.2 | 8 | 1637 | AAH12742.1 | |
| BC014397.1 | 8 | 1637 | AAH14397.1 | |
| BC093049.1 | 6 | 1223 | ||
| BI092736.1 | 3 | 575 | ||
| BX340370.2 | 2 | 425 | ||
| CR936682.1 | 6 | 1528 | ||
| CR936799.1 | 6 | 6509 | ||
| DQ266432.1 | 7 | 1516 | ABB73038.1 | |
| L42176.1 | 7 | 1196 | AAA85333.1 | |
| U29332.1 | 6 | 1179 | AAC52073.1 | |
| U60117.1 | AAC50794.1 |
More Information
Set Membership: |
|

Genomic Map