Assay Details
Target Gene Details
Entrez Gene ID: | 3817 |
Gene Name: | kallikrein related peptidase 2 |
Gene Aliases: |
KLK2A2, hGK-1, hK2 |
Location: |
Chr.19:50873433-50880567 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK2 | NM_001002231.2 | NP_001002231.1 | ||
| NM_001256080.1 | NP_001243009.1 | |||
| NM_005551.4 | NP_005542.1 | |||
| NR_045762.1 | ||||
| NR_045763.1 | 2 | 228 | ||
| AF188745.1 | AAF08275.1 | |||
| AF188746.1 | AAF08276.1 | |||
| AF188747.1 | AAF08277.1 | |||
| AK300527.1 | 2 | 228 | ||
| AK300549.1 | ||||
| AK300566.1 | ||||
| AK309969.1 | ||||
| AK309973.1 | ||||
| AK309982.1 | ||||
| AK309985.1 | ||||
| AK315890.1 | ||||
| AY429509.1 | AAR10468.1 | |||
| AY429510.1 | AAR10469.1 | |||
| BC005196.1 | AAH05196.1 | |||
| BT006650.1 | AAP35296.1 | |||
| DA865689.1 | ||||
| S39329.1 | AAD13816.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2531 | Chr.19:50830372 - 50875341 on Build GRCh38 | Deletion |
|
| esv3644659 | Chr.19:50869934 - 50881389 on Build GRCh38 | Gain |
|
| dgv1059e212 | Chr.19:50873163 - 50880505 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map