Assay Details
Target Gene Details
Entrez Gene ID: | 1628 |
Gene Name: | D-box binding PAR bZIP transcription factor |
Gene Aliases: |
DABP |
Location: |
Chr.19:48630560-48637550 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DBP | NM_001352.4 | NP_001343.2 | ||
| XM_017026388.1 | XP_016881877.1 | |||
| AB208807.1 | 2 | 3008 | BAD92044.1 | |
| AK310025.1 | 3 | 1134 | ||
| BC011965.1 | AAH11965.1 | |||
| BT006836.1 | AAP35482.1 | |||
| CA425431.1 | ||||
| D28468.1 | BAA05833.1 | |||
| U06936.1 | AAA81374.1 | |||
| U79283.1 | AAB50219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| dgv3617n100 | Chr.19:48532114 - 48643732 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map