Assay Details
Target Gene Details
Entrez Gene ID: | 84142 |
Gene Name: | family with sequence similarity 175 member A |
Gene Aliases: |
ABRA1, CCDC98 |
Location: |
Chr.4:83459515-83485181 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM175A | NM_139076.2 | 9 | 1170 | NP_620775.2 |
| XM_005263280.4 | 10 | 1303 | XP_005263337.1 | |
| XM_017008694.1 | 8 | 1076 | XP_016864183.1 | |
| AK021582.1 | 4 | 1378 | ||
| AK022704.1 | 9 | 1156 | BAB14189.1 | |
| AK023676.1 | 8 | 1070 | BAB14635.1 | |
| AY358576.1 | 9 | 1121 | AAQ88939.1 | |
| BC039573.1 | 9 | 1117 | AAH39573.1 | |
| EF531340.1 | 9 | 1105 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893971 | Chr.4:83096792 - 83493129 on Build GRCh38 | Gain |
|
| esv3601197 | Chr.4:83448066 - 83468701 on Build GRCh38 | Loss |
|
| esv3601196 | Chr.4:83419958 - 83470239 on Build GRCh38 | Loss |
|
| nsv829994 | Chr.4:83445146 - 83637163 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map