Assay Details
Target Gene Details
Entrez Gene ID: | 135112 |
Gene Name: | nuclear receptor coactivator 7 |
Gene Aliases: |
ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3 |
Location: |
Chr.6:125780948-125932030 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCOA7 | NM_001199619.1 | NP_001186548.1 | ||
| NM_001199620.1 | NP_001186549.1 | |||
| XM_006715340.3 | 1 | 41 | XP_006715403.1 | |
| XM_017010270.1 | XP_016865759.1 | |||
| XM_017010274.1 | 1 | 203 | XP_016865763.1 | |
| AF493978.1 | AAM27392.1 | |||
| AK307395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950505 | Chr.6:125772255 - 125803354 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map