Assay Details
Target Gene Details
Entrez Gene ID: | 6004 |
Gene Name: | regulator of G-protein signaling 16 |
Gene Aliases: |
A28-RGS14, A28-RGS14P, RGS-R |
Location: |
Chr.1:182598623-182604413 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS16 | NM_002928.3 | 5 | 1341 | NP_002919.3 |
| U70426.1 | 5 | 1279 | AAC16912.1 | |
| U94829.1 | 5 | 1190 | AAC52040.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825531 | Chr.1:182473650 - 182643170 on Build GRCh38 | Loss |
|
| nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain |
|
| esv1010175 | Chr.1:182573456 - 182616228 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map