Assay Details
Target Gene Details
Entrez Gene ID: | 55561 |
Gene Name: | CDC42 binding protein kinase gamma |
Gene Aliases: |
DMPK2, HSMDPKIN, KAPPA-200, MRCKG, MRCKgamma |
Location: |
Chr.11:64823809-64844685 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC42BPG | NM_017525.2 | NP_059995.2 | ||
| XM_011545155.2 | XP_011543457.1 | |||
| XM_011545156.2 | XP_011543458.1 | |||
| XM_011545157.2 | XP_011543459.1 | |||
| XM_011545158.2 | XP_011543460.1 | |||
| XM_011545159.2 | XP_011543461.1 | |||
| XM_011545160.2 | XP_011543462.1 | |||
| XM_011545161.2 | XP_011543463.1 | |||
| XM_017017996.1 | XP_016873485.1 | |||
| XM_017017997.1 | XP_016873486.1 | |||
| XM_017017998.1 | XP_016873487.1 | |||
| XM_017017999.1 | XP_016873488.1 | |||
| AY648038.1 | AAT67172.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555207 | Chr.11:64832530 - 64853046 on Build GRCh38 | Loss |
|
| nsv528506 | Chr.11:64577976 - 64933558 on Build GRCh38 | Loss |
|
| nsv469962 | Chr.11:64759608 - 64854101 on Build GRCh38 | Loss |
|
| nsv951013 | Chr.11:64777329 - 64847128 on Build GRCh38 | Deletion |
|
| nsv832189 | Chr.11:64719711 - 64852727 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map