Assay Details
Target Gene Details
Entrez Gene ID: | 84816 |
Gene Name: | reticulon 4 interacting protein 1 |
Gene Aliases: |
NIMP, OPA10 |
Location: |
Chr.6:106559237-106630921 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTN4IP1 | NM_001318746.1 | 9 | 1053 | NP_001305675.1 |
| NM_032730.5 | 9 | 1639 | NP_116119.2 | |
| XM_011536192.2 | XP_011534494.1 | |||
| XM_017011375.1 | XP_016866864.1 | |||
| XM_017011376.1 | XP_016866865.1 | |||
| AF336861.1 | AAK64603.1 | |||
| AF439711.1 | AAL34525.1 | |||
| AK095207.1 | 9 | 1053 | BAC04499.1 | |
| AK313962.1 | ||||
| AY063761.1 | AAL40856.1 | |||
| BC006399.2 | AAH06399.2 |
More Information
Set Membership: |
|

Genomic Map