Assay Details
Target Gene Details
Entrez Gene ID: | 8905 |
Gene Name: | adaptor related protein complex 1 sigma 2 subunit |
Gene Aliases: |
DC22, MRX59, MRXS21, MRXS5, MRXSF, PGS, SIGMA1B |
Location: |
Chr.X:15825806-15855014 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP1S2 | NM_001272071.1 | NP_001259000.1 | ||
| NM_003916.4 | NP_003907.3 | |||
| BI462192.1 | ||||
| DC401898.1 |
More Information
Set Membership: |
|

Genomic Map