Assay Details
Target Gene Details
Entrez Gene ID: | 221833 |
Gene Name: | Sp8 transcription factor |
Gene Aliases: |
BTD |
Location: |
Chr.7:20782274-20786889 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP8 | NM_182700.5 | 2 | 1812 | NP_874359.2 |
| NM_198956.3 | 3 | 1908 | NP_945194.1 | |
| AK056857.1 | 3 | 1641 | BAB71297.1 | |
| AK290465.1 | 3 | 1686 | ||
| AK299947.1 | 3 | 1583 | ||
| BC038669.1 | 3 | 1683 | AAH38669.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv824024 | Chr.7:20769944 - 20804515 on Build GRCh38 | Gain |
|
| nsv1030726 | Chr.7:20466472 - 21047100 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map