Assay Details
Target Gene Details
Entrez Gene ID: | 8567 |
Gene Name: | MAP kinase activating death domain |
Gene Aliases: |
DENN, IG20, RAB3GEP |
Location: |
Chr.11:47269376-47330031 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MADD | NM_001135944.1 | NP_001129416.1 | ||
| NM_003682.3 | NP_003673.3 | |||
| NM_130470.2 | NP_569826.2 | |||
| NM_130471.2 | NP_569827.2 | |||
| NM_130472.2 | NP_569828.2 | |||
| NM_130473.2 | NP_569829.2 | |||
| NM_130474.2 | NP_569830.2 | |||
| XM_017018485.1 | XP_016873974.1 | |||
| XM_017018489.1 | XP_016873978.1 | |||
| XM_017018497.1 | XP_016873986.1 | |||
| XM_017018499.1 | XP_016873988.1 | |||
| XM_017018502.1 | XP_016873991.1 | |||
| XM_017018504.1 | XP_016873993.1 | |||
| XM_017018506.1 | XP_016873995.1 | |||
| AF440100.1 | AAL40265.1 | |||
| AF440101.1 | AAL40266.1 | |||
| AF440102.1 | AAL40267.1 | |||
| AF440103.1 | AAL40268.1 | |||
| AF440434.1 | AAL35261.1 | |||
| AK292442.1 | ||||
| AU077339.1 | ||||
| DA187733.1 | ||||
| U77352.1 | AAB57735.1 |
More Information
Set Membership: |
|

Genomic Map