Assay Details
Target Gene Details
Entrez Gene ID: | 6801 |
Gene Name: | striatin |
Gene Aliases: |
PPP2R6A, SG2NA |
Location: |
Chr.2:36837698-36966607 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STRN | NM_003162.3 | 18 | 3064 | NP_003153.2 |
| XM_005264519.4 | 17 | 3088 | XP_005264576.1 | |
| XM_011533073.2 | 19 | 3286 | XP_011531375.1 | |
| AK026934.1 | 15 | 2635 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3790n100 | Chr.2:36800676 - 36868752 on Build GRCh38 | Gain |
|
| nsv428395 | Chr.2:36763205 - 37073209 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map