Assay Details
Target Gene Details
Entrez Gene ID: | 146894 |
Gene Name: | CD300 molecule like family member g |
Gene Aliases: |
CLM-9, CLM9, NEPMUCIN, TREM-4, TREM4 |
Location: |
Chr.17:43846777-43863639 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD300LG | NM_145273.3 | 7 | 1339 | NP_660316.2 |
| XM_005257075.2 | 8 | 1677 | XP_005257132.1 | |
| XM_005257076.2 | 7 | 1575 | XP_005257133.1 | |
| XM_006721721.2 | 8 | 1830 | XP_006721784.1 | |
| XM_006721722.2 | 7 | 1728 | XP_006721785.1 | |
| XM_017024235.1 | 6 | 1455 | XP_016879724.1 | |
| AY358364.1 | 7 | 1294 | AAQ88730.1 | |
| BC025395.2 | 7 | 1311 | AAH25395.1 |
More Information
Set Membership: |
|

Genomic Map