Assay Details
Target Gene Details
Entrez Gene ID: | 338442 |
Gene Name: | hydroxycarboxylic acid receptor 2 |
Gene Aliases: |
GPR109A, HCA2, HM74a, HM74b, NIACR1, PUMAG, Puma-g |
Location: |
Chr.12:122701293-122703357 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCAR2 | NM_177551.3 | 1 | 316 | NP_808219.1 |
| AK290915.1 | 1 | 255 | ||
| AL698846.1 | 1 | 316 | ||
| AY148884.1 | 1 | 302 | AAN71621.1 | |
| BC027965.1 | 1 | 302 | AAH27965.1 | |
| BC056419.1 | 1 | 288 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26589 | Chr.12:122693559 - 122726124 on Build GRCh38 | Gain |
|
| esv8582 | Chr.12:122701904 - 122715580 on Build GRCh38 | Loss |
|
| nsv973132 | Chr.12:122700236 - 122716990 on Build GRCh38 | Duplication |
|
| esv8843 | Chr.12:122697044 - 122710672 on Build GRCh38 | Loss |
|
| dgv308e214 | Chr.12:122701033 - 122717012 on Build GRCh38 | Loss |
|
| esv2746500 | Chr.12:122696739 - 122711304 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map