Assay Details
Target Gene Details
Entrez Gene ID: | 3981 |
Gene Name: | DNA ligase 4 |
Gene Aliases: |
LIG4S |
Location: |
Chr.13:108207442-108218368 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIG4 | NM_001098268.1 | 2 | 1054 | NP_001091738.1 |
| NM_002312.3 | 2 | 1092 | NP_002303.2 | |
| NM_206937.1 | 3 | 971 | NP_996820.1 | |
| XM_005254056.1 | 3 | 936 | XP_005254113.1 | |
| XM_005254057.4 | 3 | 1106 | XP_005254114.1 | |
| XM_005254058.3 | 2 | 2552 | XP_005254115.1 | |
| XM_006719951.3 | 3 | 2811 | XP_006720014.1 | |
| XM_006719952.1 | 4 | 1404 | XP_006720015.1 | |
| XM_011521091.2 | 4 | 1433 | XP_011519393.1 | |
| XM_011521092.2 | 3 | 1009 | XP_011519394.1 | |
| XM_017020563.1 | 3 | 2653 | XP_016876052.1 | |
| XM_017020564.1 | 2 | 1079 | XP_016876053.1 | |
| XM_017020565.1 | 4 | 1450 | XP_016876054.1 | |
| XM_017020566.1 | 4 | 1353 | XP_016876055.1 | |
| XM_017020567.1 | 3 | 1007 | XP_016876056.1 | |
| XM_017020568.1 | 3 | 2629 | XP_016876057.1 | |
| XM_017020569.1 | 4 | 3074 | XP_016876058.1 | |
| XM_017020570.1 | 4 | 1353 | XP_016876059.1 | |
| XM_017020571.1 | 4 | 3158 | XP_016876060.1 | |
| XM_017020572.1 | 3 | 2545 | XP_016876061.1 | |
| XM_017020573.1 | 2 | 855 | XP_016876062.1 | |
| AK292419.1 | 3 | 880 | ||
| AK314844.1 | 3 | 936 | ||
| BC037491.2 | 3 | 971 | AAH37491.1 | |
| BX648833.1 | 2 | 1055 | ||
| X83441.1 | 2 | 1092 | CAA58467.1 |
More Information
Set Membership: |
|

Genomic Map