Assay Details
Target Gene Details
Entrez Gene ID: | 6810 |
Gene Name: | syntaxin 4 |
Gene Aliases: |
STX4A, p35-2 |
Location: |
Chr.16:31033095-31040168 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STX4 | NM_001272095.1 | NP_001259024.1 | ||
| NM_001272096.1 | NP_001259025.1 | |||
| NM_004604.4 | NP_004595.2 | |||
| XM_005255521.1 | XP_005255578.1 | |||
| XM_011545925.2 | XP_011544227.1 | |||
| AF026007.1 | AAB88810.1 | |||
| AF318489.1 | AAG40313.1 | |||
| AJ000541.1 | CAA04174.1 | |||
| AK091833.1 | ||||
| AK296652.1 | ||||
| AK315716.1 | ||||
| BC002436.2 | AAH02436.1 | |||
| BQ775670.1 | ||||
| BT007326.1 | AAP35990.1 | |||
| CR541806.1 | CAG46605.1 | |||
| U07158.1 | AAA20967.1 | |||
| X85784.1 | CAA59769.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457483 | Chr.16:30892358 - 31180161 on Build GRCh38 | Loss |
|
| nsv833193 | Chr.16:30949475 - 31122283 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map