Assay Details
Target Gene Details
Entrez Gene ID: | 79982 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B14 |
Gene Aliases: |
EGNR9427, PRO34683 |
Location: |
Chr.4:99896248-99946726 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB14 | NM_001031723.3 | NP_001026893.1 | ||
| NM_001278310.1 | NP_001265239.1 | |||
| XM_011532262.2 | XP_011530564.1 | |||
| XM_011532263.2 | XP_011530565.1 | |||
| XM_011532265.2 | XP_011530567.1 | |||
| XM_017008628.1 | XP_016864117.1 | |||
| AK023700.1 | BAB14645.1 | |||
| AK024343.1 | BAB14893.1 | |||
| AL833076.2 | CAD89928.1 | |||
| AL833412.1 | ||||
| AY186740.1 | AAO31693.1 | |||
| AY358272.1 | AAQ88639.1 | |||
| BC022248.1 | AAH22248.1 |

Genomic Map