Assay Details
Target Gene Details
Entrez Gene ID: | 124923 |
Gene Name: | uncharacterized serine/threonine-protein kinase SgK494 |
Gene Aliases: |
- |
Location: |
Chr.17:28607964-28614193 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGK494 | NM_001174103.1 | 12 | 3451 | NP_001167574.1 |
| AK056265.1 | 1 | 1260 | ||
| AK057735.1 | 10 | 1105 | BAB71555.1 | |
| AK294189.1 | ||||
| BC045622.1 | AAH45622.1 |
Target Gene Details
Entrez Gene ID: | 100506436 |
Gene Name: | SPAG5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:28599133-28617377 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPAG5-AS1 | NR_040012.1 | |||
| DB457167.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482200 | Chr.17:28607964 - 28614193 on Build GRCh38 | Loss |
|
| nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map