Assay Details
Target Gene Details
Entrez Gene ID: | 1453 |
Gene Name: | casein kinase 1 delta |
Gene Aliases: |
ASPS, CKIdelta, FASPS2, HCKID |
Location: |
Chr.17:82239317-82273742 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSNK1D | NM_001893.4 | 9 | 1881 | NP_001884.2 |
| NM_139062.2 | 10 | 1945 | NP_620693.1 | |
| NR_110578.1 | 9 | 1740 | ||
| XM_005256337.4 | XP_005256394.1 | |||
| AB091044.1 | 9 | 1593 | BAC10903.1 | |
| AB209463.1 | 8 | 4281 | BAD92700.1 | |
| AK096950.1 | 2 | 909 | ||
| BC003558.1 | 9 | 1852 | AAH03558.1 | |
| BC015775.1 | 10 | 1924 | AAH15775.1 | |
| U29171.1 | 9 | 1721 | AAC50807.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain |
|
| nsv828138 | Chr.17:82244143 - 82244825 on Build GRCh38 | Gain |
|
| nsv833572 | Chr.17:82204977 - 82388835 on Build GRCh38 | Loss |
|
| esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map