Assay Details
Target Gene Details
Entrez Gene ID: | 23299 |
Gene Name: | BICD cargo adaptor 2 |
Gene Aliases: |
SMALED2, bA526D8.1 |
Location: |
Chr.9:92711363-92764841 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BICD2 | NM_001003800.1 | 7 | 4381 | NP_001003800.1 |
| NM_015250.3 | 8 | 2590 | NP_056065.1 | |
| XM_017014551.1 | 8 | 2711 | XP_016870040.1 | |
| AB014599.1 | 7 | 4301 | BAA31674.1 | |
| AY052562.1 | 8 | 2590 | AAL12246.1 | |
| BC004296.1 | 5 | 1821 | AAH04296.1 | |
| BC010428.1 | 1 | 576 | AAH10428.1 | |
| BC073970.1 | 7 | 4396 | AAH73970.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621086 | Chr.9:92660561 - 92718718 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map