Assay Details
Target Gene Details
Entrez Gene ID: | 140876 |
Gene Name: | family with sequence similarity 65 member C |
Gene Aliases: |
C20orf175, C20orf176 |
Location: |
Chr.20:50586108-50692004 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM65C | NM_001290268.1 | 22 | 3789 | NP_001277197.1 |
| NM_080829.3 | 22 | 3722 | NP_543019.2 | |
| NR_110890.1 | 22 | 3948 | ||
| XM_005260294.3 | 23 | 3786 | XP_005260351.1 | |
| XM_006723713.3 | 22 | 3834 | XP_006723776.1 | |
| XM_011528578.2 | 23 | 3814 | XP_011526880.1 | |
| XM_011528579.2 | 23 | 3952 | XP_011526881.1 | |
| XM_011528580.2 | 23 | 4006 | XP_011526882.1 | |
| XM_011528581.2 | 23 | 3990 | XP_011526883.1 | |
| XM_011528585.2 | 12 | 3137 | XP_011526887.1 | |
| XM_017027681.1 | 14 | 2782 | XP_016883170.1 | |
| AI056877.1 | ||||
| AV703941.1 | 1 | 91 | ||
| BQ446237.1 | 1 | 605 | ||
| BX111440.1 | 1 | 408 | ||
| CD558804.1 |
More Information
Set Membership: |
|

Genomic Map