Assay Details
Target Gene Details
Entrez Gene ID: | 51348 |
Gene Name: | killer cell lectin like receptor F1 |
Gene Aliases: |
CLEC5C, NKp80 |
Location: |
Chr.12:9827303-9845005 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLRF1 | NM_001291822.1 | NP_001278751.1 | ||
| NM_001291823.1 | NP_001278752.1 | |||
| NM_016523.2 | NP_057607.1 | |||
| NR_120305.1 | ||||
| XM_017019414.1 | XP_016874903.1 | |||
| XM_017019415.1 | XP_016874904.1 | |||
| AF175206.1 | AAF37804.1 | |||
| AF175207.1 | AAF37805.1 | |||
| AF267244.1 | AAL14873.1 | |||
| AF267245.1 | AAL14874.1 | |||
| AJ305370.1 | CAC29425.1 | |||
| BC096734.1 | AAH96734.1 | |||
| BC098166.1 | AAH98166.1 | |||
| BC098257.1 | AAH98257.1 | |||
| BC098354.1 | AAH98354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map