Assay Details
Target Gene Details
Entrez Gene ID: | 90338 |
Gene Name: | zinc finger protein 160 |
Gene Aliases: |
F11, HKr18, HZF5, KR18 |
Location: |
Chr.19:53064882-53103434 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF160 | NM_001102603.1 | 7 | 3824 | NP_001096073.1 |
| NM_001322128.1 | 7 | 3832 | NP_001309057.1 | |
| NM_001322129.1 | 6 | 3927 | NP_001309058.1 | |
| NM_001322130.1 | 7 | 4056 | NP_001309059.1 | |
| NM_001322131.1 | 6 | 3931 | NP_001309060.1 | |
| NM_001322132.1 | 7 | 3820 | NP_001309061.1 | |
| NM_001322133.1 | 7 | 3844 | NP_001309062.1 | |
| NM_001322134.1 | 7 | 3848 | NP_001309063.1 | |
| NM_001322135.1 | 6 | 3703 | NP_001309064.1 | |
| NM_001322136.1 | 5 | 3560 | NP_001309065.1 | |
| NM_001322137.1 | 8 | 3880 | NP_001309066.1 | |
| NM_001322138.1 | 7 | 3763 | NP_001309067.1 | |
| NM_001322139.1 | 9 | 3977 | NP_001309068.1 | |
| NM_033288.3 | 6 | 3707 | NP_150630.2 | |
| NM_198893.2 | 7 | 3792 | NP_942596.1 | |
| XM_017027445.1 | 4 | 4056 | XP_016882934.1 | |
| XM_017027446.1 | 6 | 4017 | XP_016882935.1 | |
| AB046831.1 | 3 | 4177 | BAB13437.1 | |
| AK024442.1 | 1 | 3071 | BAB15732.1 | |
| BC094880.1 | 7 | 3709 | AAH94880.1 | |
| BF570224.1 | 1 | 279 | ||
| CR749613.1 | 1 | 2522 | CAH18407.1 | |
| X77744.1 | 1 | 1710 | ||
| X78928.1 | CAA55528.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2718787 | Chr.19:52570219 - 53366454 on Build GRCh38 | Deletion |
|
| nsv428370 | Chr.19:52907206 - 53149322 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map