Assay Details
Target Gene Details
Entrez Gene ID: | 114780 |
Gene Name: | polycystin 1 like 2 (gene/pseudogene) |
Gene Aliases: |
PC1L2 |
Location: |
Chr.16:81100879-81220370 on Build GRCh38 |
Assay Gene Location: | Within Exon 44 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKD1L2 | NM_001278425.1 | 32 | 5602 | NP_001265354.1 |
| NM_052892.3 | 43 | 7508 | NP_443124.3 | |
| NR_126532.1 | 43 | 7507 | ||
| AK131378.1 | 11 | 1896 | BAD18529.1 | |
| AL832135.1 | 3 | 5211 | ||
| AW082870.1 | 1 | 115 | ||
| AY371495.1 | 32 | 5564 | AAQ73173.1 | |
| DB306292.1 | 1 | 117 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751615 | Chr.16:81059494 - 81273994 on Build GRCh38 | Gain |
|
| nsv1062724 | Chr.16:81018707 - 81103402 on Build GRCh38 | Gain |
|
| nsv1067144 | Chr.16:81095693 - 81148036 on Build GRCh38 | Loss |
|
| esv2422360 | Chr.16:81032428 - 81147122 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80096476] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map