Assay Details
Target Gene Details
Entrez Gene ID: | 151112 |
Gene Name: | zinc finger SWIM-type containing 2 |
Gene Aliases: |
MEX, ZZZ2 |
Location: |
Chr.2:186827480-186849170 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZSWIM2 | NM_182521.2 | 9 | 1230 | NP_872327.2 |
| AK128006.1 | 9 | 1230 | ||
| BC031094.1 | 9 | 1229 | AAH31094.1 |
More Information
Set Membership: |
|

Genomic Map