Assay Details
Target Gene Details
Entrez Gene ID: | 202243 |
Gene Name: | coiled-coil domain containing 125 |
Gene Aliases: |
KENAE |
Location: |
Chr.5:69273087-69333069 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC125 | NM_001297696.1 | 11 | 2583 | NP_001284625.1 |
| NM_001297697.1 | 13 | 2639 | NP_001284626.1 | |
| NM_176816.4 | 12 | 2800 | NP_789786.2 | |
| XM_005248458.4 | 12 | 3037 | XP_005248515.1 | |
| XM_005248459.4 | 12 | 2839 | XP_005248516.1 | |
| XM_005248461.3 | 11 | 2526 | XP_005248518.1 | |
| XM_005248463.3 | 10 | 2394 | XP_005248520.1 | |
| XM_006714569.3 | 12 | 2827 | XP_006714632.1 | |
| XM_006714570.3 | 12 | 2611 | XP_006714633.1 | |
| XM_011543255.2 | 12 | 2817 | XP_011541557.1 | |
| XM_011543256.2 | 12 | 2818 | XP_011541558.1 | |
| XM_011543257.2 | 11 | 2734 | XP_011541559.1 | |
| XM_011543258.2 | XP_011541560.1 | |||
| XM_011543259.2 | XP_011541561.1 | |||
| XM_017009206.1 | 12 | 3032 | XP_016864695.1 | |
| XM_017009207.1 | XP_016864696.1 | |||
| XM_017009208.1 | 10 | 2666 | XP_016864697.1 | |
| XM_017009209.1 | 11 | 2664 | XP_016864698.1 | |
| XM_017009210.1 | 9 | 2587 | XP_016864699.1 | |
| XM_017009211.1 | 6 | 2069 | XP_016864700.1 | |
| AB024691.1 | 13 | 2453 | BAC57450.1 | |
| AB024692.1 | 12 | 2156 | BAC57451.1 | |
| BC113749.1 | 6 | 2052 | AAI13750.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5694n100 | Chr.5:69081291 - 69374274 on Build GRCh38 | Gain |
|
| nsv821643 | Chr.5:69272838 - 71076658 on Build GRCh38 | Loss |
|

Genomic Map