Assay Details
Target Gene Details
Entrez Gene ID: | 51004 |
Gene Name: | coenzyme Q6, monooxygenase |
Gene Aliases: |
CGI-10, CGI10, COQ10D6 |
Location: |
Chr.14:73949934-73963112 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COQ6 | NM_182476.2 | 2 | 312 | NP_872282.1 |
| NM_182480.2 | 2 | 315 | NP_872286.2 | |
| XM_006720156.1 | 2 | 356 | XP_006720219.1 | |
| XM_011536807.1 | 2 | 284 | XP_011535109.1 | |
| XM_011536808.1 | 2 | 162 | XP_011535110.1 | |
| XM_011536809.2 | 2 | 309 | XP_011535111.1 | |
| XM_011536810.2 | 2 | 284 | XP_011535112.1 | |
| XM_017021351.1 | 2 | 133 | XP_016876840.1 | |
| XM_017021352.1 | 2 | 254 | XP_016876841.1 | |
| AF132944.1 | 2 | 238 | AAD27719.1 | |
| AK130167.1 | 2 | 269 | ||
| AK222965.1 | 2 | 254 | BAD96685.1 | |
| AK294370.1 | 2 | 254 | ||
| AK295517.1 | 2 | 254 | ||
| AK296040.1 | 2 | 323 | ||
| AK296386.1 | 2 | 254 | ||
| AK303304.1 | 2 | 263 | ||
| AK303350.1 | 2 | 134 | ||
| AK307306.1 | 2 | 356 | ||
| AK316356.1 | 2 | 139 | ||
| AL521052.3 | 2 | 343 | ||
| AL557147.3 | 2 | 304 | ||
| BC014181.2 | 2 | 261 | AAH14181.1 | |
| BC014483.1 | 2 | 249 | AAH14483.1 | |
| BX248000.1 | 2 | 309 | CAD62332.1 | |
| BX342249.2 | 2 | 313 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1352 | Chr.14:73951320 - 73996025 on Build GRCh38 | Deletion |
|
| nsv1053281 | Chr.14:73518500 - 74050155 on Build GRCh38 | Gain |
|
| dgv1937n100 | Chr.14:73950133 - 74115803 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map