Assay Details
Target Gene Details
Entrez Gene ID: | 6659 |
Gene Name: | SRY-box 4 |
Gene Aliases: |
EVI16 |
Location: |
Chr.6:21593741-21598619 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX4 | NM_003107.2 | 1 | 3455 | NP_003098.1 |
| BM459088.1 | 1 | 355 | ||
| CF147018.1 | ||||
| HY065060.1 | 1 | 344 |
More Information
Set Membership: |
|

Genomic Map