Assay Details
Target Gene Details
Entrez Gene ID: | 136991 |
Gene Name: | ankyrin repeat, SAM and basic leucine zipper domain containing 1 |
Gene Aliases: |
ALP1, ANKL1, C7orf7, CT1.19, GASZ, Orf3 |
Location: |
Chr.7:117363222-117427523 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASZ1 | NM_001301821.1 | 10 | 1050 | NP_001288750.1 |
| NM_001301822.1 | 9 | 938 | NP_001288751.1 | |
| NM_130768.2 | 10 | 1050 | NP_570124.1 | |
| AF461259.1 | 10 | 1016 | AAL68815.1 | |
| AK093445.1 | 10 | 1003 | BAC04167.1 | |
| BC034963.1 | 10 | 1025 | AAH34963.1 | |
| BC126186.1 | 10 | 1009 | ||
| BC126188.1 | 10 | 1009 | ||
| BC144212.1 | 10 | 1009 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608256 | Chr.7:117288317 - 117400063 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map