Assay Details
Target Gene Details
Entrez Gene ID: | 64816 |
Gene Name: | cytochrome P450 family 3 subfamily A member 43 |
Gene Aliases: |
- |
Location: |
Chr.7:99828013-99867138 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP3A43 | NM_001278921.1 | NP_001265850.1 | ||
| NM_022820.4 | NP_073731.1 | |||
| NM_057095.2 | NP_476436.1 | |||
| NM_057096.3 | NP_476437.1 | |||
| NR_103868.1 | ||||
| NR_103869.1 | ||||
| AF280108.1 | AAG33009.1 | |||
| AF280109.1 | AAG33010.1 | |||
| AF280110.1 | AAG33011.1 | |||
| AF280111.1 | AAG33012.1 | |||
| AF280112.1 | AAG33013.1 | |||
| AF280113.1 | AAG33014.1 | |||
| AF319634.1 | AAK00325.1 | |||
| AF337813.1 | AAK38841.1 | |||
| AY390423.1 | AAQ92351.1 | |||
| AY390424.1 | AAQ92352.1 | |||
| AY390425.1 | AAQ92353.1 | |||
| AY390426.1 | AAQ92354.1 | |||
| BC100981.3 | AAI00982.1 | |||
| BC100982.3 | AAI00983.1 |
More Information
Set Membership: |
|

Genomic Map