Assay Details
Target Gene Details
Entrez Gene ID: | 3737 |
Gene Name: | potassium voltage-gated channel subfamily A member 2 |
Gene Aliases: |
EIEE32, HBK5, HK4, HUKIV, KV1.2, MK2, NGK1, RBK2 |
Location: |
Chr.1:110593580-110631536 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNA2 | NM_001204269.1 | NP_001191198.1 | ||
| NM_004974.3 | 3 | 1685 | NP_004965.1 | |
| XM_011541396.2 | 3 | 1679 | XP_011539698.1 | |
| XM_011541397.2 | 4 | 1960 | XP_011539699.1 | |
| XM_011541398.2 | 3 | 1727 | XP_011539700.1 | |
| XM_011541399.2 | 3 | 1768 | XP_011539701.1 | |
| XM_011541400.2 | 3 | 1698 | XP_011539702.1 | |
| XM_017001213.1 | 3 | 1660 | XP_016856702.1 | |
| AK290061.1 | 3 | 1685 | ||
| BC043564.1 | AAH43564.1 | |||
| BM682747.1 | ||||
| L02752.1 | 1 | 1064 | AAA36141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv547499 | Chr.1:110581114 - 110618388 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map