Assay Details
Target Gene Details
Entrez Gene ID: | 25822 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B5 |
Gene Aliases: |
Hsc40 |
Location: |
Chr.9:34989369-34998433 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB5 | NM_001135004.2 | NP_001128476.2 | ||
| NM_001135005.2 | NP_001128477.1 | |||
| NM_012266.5 | NP_036398.3 | |||
| XM_005251428.3 | XP_005251485.1 | |||
| XM_006716751.2 | XP_006716814.1 | |||
| XM_011517842.2 | XP_011516144.1 | |||
| AF087870.1 | AAM10498.1 | |||
| AF088982.1 | AAC35860.1 | |||
| AK023253.1 | ||||
| AK299647.1 | ||||
| AK314671.1 | ||||
| AL536638.3 | ||||
| BI915181.1 | ||||
| BU845478.1 | ||||
| BX419166.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6527 | Chr.9:34984490 - 35015049 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map