Assay Details
Target Gene Details
Entrez Gene ID: | 51562 |
Gene Name: | MAP3K12 binding inhibitory protein 1 |
Gene Aliases: |
- |
Location: |
Chr.14:36298558-36320676 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBIP | NM_001144891.1 | 9 | 1391 | NP_001138363.1 |
| NM_001308110.1 | 8 | 1296 | NP_001295039.1 | |
| NM_016586.2 | 9 | 1394 | NP_057670.2 | |
| XM_005267754.3 | 8 | 1323 | XP_005267811.1 | |
| XM_005267756.4 | 8 | 1288 | XP_005267813.1 | |
| XM_017021367.1 | 7 | 1222 | XP_016876856.1 | |
| AA761503.1 | 1 | 268 | ||
| AB038523.1 | 9 | 1363 | BAA94083.1 | |
| AF208857.1 | 10 | 1562 | AAF64271.1 | |
| AK293473.1 | 9 | 1275 | ||
| BC005197.1 | 9 | 1353 | AAH05197.1 | |
| BC016821.1 | 9 | 1346 | AAH16821.1 | |
| BX248287.1 | 8 | 1216 | CAD62615.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv83e55 | Chr.14:36267143 - 36313743 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map