Assay Details
Target Gene Details
Entrez Gene ID: | 51 |
Gene Name: | acyl-CoA oxidase 1 |
Gene Aliases: |
ACOX, PALMCOX, SCOX |
Location: |
Chr.17:75941507-75979434 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACOX1 | NM_001185039.1 | 14 | 4880 | NP_001171968.1 |
| NM_004035.6 | 14 | 4706 | NP_004026.2 | |
| NM_007292.5 | 14 | 4706 | NP_009223.2 | |
| XM_011524868.2 | 15 | 4865 | XP_011523170.1 | |
| XM_011524869.2 | 13 | 4543 | XP_011523171.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3641270 | Chr.17:75929772 - 75959761 on Build GRCh38 | Gain |
|
| esv3641271 | Chr.17:75936747 - 75955938 on Build GRCh38 | Gain |
|
| nsv833545 | Chr.17:75738140 - 75945931 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map