Assay Details
Target Gene Details
Entrez Gene ID: | 8550 |
Gene Name: | mitogen-activated protein kinase-activated protein kinase 5 |
Gene Aliases: |
MAPKAP-K5, MK-5, MK5, PRAK |
Location: |
Chr.12:111842228-111902225 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPKAPK5 | NM_003668.3 | 14 | 2169 | NP_003659.2 |
| NM_139078.2 | 14 | 2175 | NP_620777.1 | |
| XM_017020125.1 | 14 | 1661 | XP_016875614.1 | |
| XM_017020126.1 | 12 | 1575 | XP_016875615.1 | |
| XM_017020127.1 | XP_016875616.1 | |||
| XM_017020128.1 | XP_016875617.1 | |||
| XM_017020129.1 | 13 | 1837 | XP_016875618.1 | |
| XM_017020130.1 | 13 | 1605 | XP_016875619.1 | |
| XM_017020131.1 | 13 | 1994 | XP_016875620.1 | |
| XM_017020132.1 | XP_016875621.1 | |||
| XM_017020133.1 | XP_016875622.1 | |||
| XM_017020134.1 | XP_016875623.1 | |||
| XM_017020135.1 | XP_016875624.1 | |||
| XM_017020136.1 | XP_016875625.1 | |||
| XM_017020137.1 | 12 | 1723 | XP_016875626.1 | |
| XM_017020138.1 | XP_016875627.1 | |||
| XM_017020139.1 | 13 | 1787 | XP_016875628.1 | |
| AK122767.1 | 14 | 2419 | ||
| AL110301.1 | 9 | 1271 | CAB53747.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044606 | Chr.12:111885161 - 111989037 on Build GRCh38 | Gain |
|
| nsv1045488 | Chr.12:111883935 - 111922897 on Build GRCh38 | Gain |
|
| esv1010936 | Chr.12:111890837 - 111902032 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map