Assay Details
Target Gene Details
Entrez Gene ID: | 2300 |
Gene Name: | forkhead box L1 |
Gene Aliases: |
FKH6, FKHL11, FREAC7 |
Location: |
Chr.16:86578509-86581698 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXL1 | NM_005250.2 | 1 | 2980 | NP_005241.1 |
| BC100027.1 | 1 | 2980 | AAI00028.1 | |
| BM675371.1 | 1 | 228 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065605 | Chr.16:86256867 - 86677986 on Build GRCh38 | Gain |
|
| dgv498n67 | Chr.16:86483168 - 86587529 on Build GRCh38 | Gain |
|
| nsv525024 | Chr.16:86479615 - 86593836 on Build GRCh38 | Loss |
|
| nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111494988] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map